The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300948.1 | NP_001287877.1 | 13 - 14 | 2426 | 72 |
NM_031490.3 | NP_113678.2 | 14 - 15 | 2558 | 72 | |
GenBank mRNA | AJ548761.1 | - | 14 - 15 | 2425 | 72 |
AK027544.1 | - | 14 - 15 | 2391 | 72 | |
AK027666.1 | - | 10 - 11 | 1593 | 72 | |
AK074775.1 | - | 14 - 15 | 2423 | 72 | |
AK127125.1 | - | 14 - 15 | 2190 | 72 | |
AK130091.1 | - | 3 - 4 | 435 | 72 | |
AK172790.1 | - | 5 - 6 | 713 | 72 | |
AL834201.1 | - | 13 - 14 | 1924 | 72 | |
BC002916.2 | - | 2 - 3 | 394 | 72 | |
BC004234.2 | - | 2 - 3 | 394 | 72 | |
BC093910.1 | - | 14 - 15 | 2392 | 72 | |
BC093912.1 | - | 14 - 15 | 2392 | 72 | |
BC110434.1 | - | 10 - 11 | 1574 | 72 | |
BC143246.1 | - | 13 - 14 | 2260 | 72 |