The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113378.1 | NP_001106849.1 | 25 - 26 | 2890 | 69 |
NM_018193.2 | NP_060663.2 | 24 - 25 | 2710 | 69 | |
XM_011521756.2 | XP_011520058.1 | 25 - 26 | 2914 | 69 | |
XM_011521757.2 | XP_011520059.1 | 25 - 26 | 2910 | 69 | |
XM_011521764.2 | XP_011520066.1 | 24 - 25 | 2734 | 69 | |
XM_011521767.2 | XP_011520069.1 | 25 - 26 | 2884 | 69 | |
GenBank mRNA | AB058697.1 | - | 11 - 12 | 2814 | 69 |
AK001581.1 | - | 18 - 19 | 2099 | 69 | |
AK027564.1 | - | 13 - 14 | 1597 | 69 | |
AK055176.1 | - | 24 - 25 | 2669 | 69 | |
BC021859.1 | - | 2 - 3 | 232 | 69 | |
BC140769.1 | - | 25 - 26 | 2847 | 69 | |
BC144483.1 | - | 25 - 26 | 2847 | 69 | |
EF469766.1 | - | 24 - 25 | 2800 | 69 | |
EF567077.1 | - | 24 - 25 | 2800 | 69 |