Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP43
Entrez Gene ID
124739
Gene Name
ubiquitin specific peptidase 43
Gene Aliases
-
Chromosome Location
Chr.17: 9643493 - 9729687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267576.1 NP_001254505.1 5 - 6 1066 88
NM_153210.4 NP_694942.3 5 - 6 1066 88
XM_011523639.2 XP_011521941.1 4 - 5 954 88
XM_011523640.1 XP_011521942.1 4 - 5 786 88
XM_011523641.1 XP_011521943.1 5 - 6 511 88
XM_011523642.2 XP_011521944.1 5 - 6 1111 88
XM_017024159.1 XP_016879648.1 4 - 5 1007 88
XM_017024160.1 XP_016879649.1 3 - 4 993 88
XM_017024161.1 XP_016879650.1 2 - 3 1079 88
XM_017024162.1 XP_016879651.1 5 - 6 1111 88
GenBank mRNA AJ583817.2 - 5 - 6 973 88
AK055188.1 - 5 - 6 547 88
BC136368.1 - 5 - 6 1066 88
BC144041.1 - 5 - 6 1066 88
BC171759.1 - 5 - 6 1066 88
BX648526.1 - 3 - 4 329 88

View Full Product Details