Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAT1
Entrez Gene ID
586
Gene Name
branched chain amino acid transaminase 1
Gene Aliases
BCATC, BCT1, ECA39, MECA39, PNAS121, PP18
Chromosome Location
Chr.12: 24810024 - 24949459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178091.1 NP_001171562.1 9 - 10 1533 95
NM_001178092.1 NP_001171563.1 8 - 9 1461 95
NM_001178093.1 NP_001171564.1 10 - 11 1240 95
NM_001178094.1 NP_001171565.1 10 - 11 1537 95
NM_005504.6 NP_005495.2 10 - 11 1644 95
XM_017019768.1 XP_016875257.1 9 - 10 1252 95
GenBank mRNA AK056255.1 - 10 - 11 1644 95
AK128527.1 - 8 - 9 1088 95
AK294879.1 - 10 - 11 1223 95
AK299088.1 - 9 - 10 1117 95
BX474651.1 - 3 - 4 290 95
BX647122.1 - 10 - 11 1246 95
CR749308.1 - 10 - 11 1446 95
U21551.1 - 9 - 10 1111 95

View Full Product Details