Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGEF1B
Entrez Gene ID
153020
Gene Name
RasGEF domain family member 1B
Gene Aliases
GPIG4
Chromosome Location
Chr.4: 81426393 - 81471928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300735.1 NP_001287664.1 3 - 4 461 78
NM_152545.2 NP_689758.1 3 - 4 461 78
XM_017007813.1 XP_016863302.1 3 - 4 549 78
GenBank mRNA AK056257.1 - 3 - 4 440 78
AK291459.1 - 3 - 4 409 78
BC036784.1 - 3 - 4 423 78
BC121003.1 - 3 - 4 376 78
BX648337.1 - 3 - 4 374 78

View Full Product Details