Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
Chromosome Location
Chr.1: 171841478 - 172418466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136127.2 NP_001129599.1 12 - 13 1674 93
NM_001278252.1 NP_001265181.1 12 - 13 1674 93
NM_015569.4 NP_056384.2 12 - 13 1674 93
XM_017000978.1 XP_016856467.1 12 - 13 1657 93
XM_017000983.1 XP_016856472.1 12 - 13 1657 93
XM_017000984.1 XP_016856473.1 12 - 13 1657 93
XM_017000985.1 XP_016856474.1 12 - 13 1657 93
GenBank mRNA AB020627.2 - 12 - 13 1625 93
AK056279.1 - 12 - 13 1654 93
AK308992.1 - 12 - 13 1623 93
AL136712.1 - 12 - 13 1652 93
BC064546.1 - 12 - 13 1515 93

View Full Product Details