The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272012.1 | NP_001258941.1 | 1 - 2 | 126 | 112 |
GenBank mRNA | AK056526.1 | - | 1 - 2 | 58 | 112 |
DA416329.1 | - | 1 - 2 | 122 | 112 |