The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022370.3 | NP_071765.2 | 9 - 10 | 1705 | 61 |
XM_017018122.1 | XP_016873611.1 | 8 - 9 | 1470 | 61 | |
GenBank mRNA | AK291045.1 | - | 9 - 10 | 1639 | 61 |
AY509035.1 | - | 9 - 10 | 1728 | 61 |