The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278529.1 | NP_001265458.1 | 3 - 4 | 1585 | 118 |
NM_018071.4 | NP_060541.3 | 3 - 4 | 1585 | 118 | |
XM_005267844.2 | XP_005267901.1 | 3 - 4 | 1621 | 118 | |
XM_011536937.2 | XP_011535239.1 | 3 - 4 | 1621 | 118 | |
XM_011536938.2 | XP_011535240.1 | 3 - 4 | 1623 | 118 | |
XM_017021434.1 | XP_016876923.1 | 4 - 5 | 1878 | 118 | |
XM_017021435.1 | XP_016876924.1 | 3 - 4 | 1622 | 118 | |
GenBank mRNA | AK024463.1 | - | 1 - 2 | 823 | 118 |
AK074057.1 | - | 3 - 4 | 1530 | 118 | |
BC142692.1 | - | 3 - 4 | 1528 | 118 | |
BC151212.1 | - | 3 - 4 | 1528 | 118 | |
BX248768.1 | - | 3 - 4 | 1572 | 118 |