The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079653.1 | NP_001073121.1 | 13 - 14 | 1983 | 79 |
NM_152744.3 | NP_689957.3 | 39 - 40 | 5838 | 79 | |
XM_011515188.1 | XP_011513490.1 | 33 - 34 | 4679 | 79 | |
XM_011515189.1 | XP_011513491.1 | 33 - 34 | 4676 | 79 | |
XM_017011837.1 | XP_016867326.1 | 33 - 34 | 4679 | 79 | |
GenBank mRNA | AK074077.1 | - | 6 - 7 | 778 | 79 |
AK074083.1 | - | 24 - 25 | 3474 | 79 | |
AL833354.1 | - | 13 - 14 | 1980 | 79 | |
AY310398.1 | - | 39 - 40 | 5838 | 79 |