Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNDC1
Entrez Gene ID
85442
Gene Name
kinase non-catalytic C-lobe domain containing 1
Gene Aliases
C10orf23, RASGEF2, bB439H18.3
Chromosome Location
Chr.10: 133160096 - 133226412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152643.6 NP_689856.6 1 - 2 100 80
XM_017016858.1 XP_016872347.1 1 - 2 468 80
XM_017016859.1 XP_016872348.1 1 - 2 471 80
GenBank mRNA AI146357.1 - 1 - 2 64 80
AK090457.1 - 1 - 2 63 80
DA388038.1 - 1 - 2 136 80

View Full Product Details