The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152643.6 | NP_689856.6 | 12 - 13 | 1909 | 87 |
XM_017016858.1 | XP_016872347.1 | 12 - 13 | 2277 | 87 | |
XM_017016859.1 | XP_016872348.1 | 12 - 13 | 2280 | 87 | |
GenBank mRNA | AB000781.1 | - | 11 - 12 | 1805 | 87 |
AB051555.1 | - | 11 - 12 | 2003 | 87 | |
AK074179.1 | - | 2 - 3 | 175 | 87 | |
AK090457.1 | - | 11 - 12 | 2816 | 87 |