Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR81
Entrez Gene ID
124997
Gene Name
WD repeat domain 81
Gene Aliases
CAMRQ2, PPP1R166, SORF-2
Chromosome Location
Chr.17: 1716523 - 1738599 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163673.1 NP_001157145.1 3 - 4 410 84
NM_001163809.1 NP_001157281.1 3 - 4 4095 84
NM_001163811.1 NP_001157283.1 3 - 4 546 84
NM_152348.3 NP_689561.2 4 - 5 1047 84
XM_011523651.2 XP_011521953.1 4 - 5 1032 84
XM_017024184.1 XP_016879673.1 3 - 4 4118 84
GenBank mRNA AK074111.1 - 3 - 4 1349 84
AK091136.1 - 4 - 5 1031 84
AK122972.1 - 3 - 4 1357 84
AK123896.1 - 3 - 4 537 84
AK127946.1 - 3 - 4 546 84
AK298567.1 - 3 - 4 410 84
AL834379.1 - 3 - 4 871 84
BC092513.1 - 3 - 4 822 84
BC114519.1 - 2 - 3 305 84
BC114568.1 - 3 - 4 372 84

View Full Product Details