Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC174
- Entrez Gene ID
-
51244
- Gene Name
- coiled-coil domain containing 174
- Gene Aliases
-
C3orf19, HSPC212, IHPM, IHPMR
- Chromosome Location
-
Chr.3: 14651683 - 14673186 on Build GRCh38
- Gene Symbol
- TMEM135
- Entrez Gene ID
-
65084
- Gene Name
- transmembrane protein 135
- Gene Aliases
-
PMP52
- Chromosome Location
-
Chr.11: 87037844 - 87328834 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC012553.1 |
- |
- |
121 |
81 |
View Full Product Details