Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC174
Entrez Gene ID
51244
Gene Name
coiled-coil domain containing 174
Gene Aliases
C3orf19, HSPC212, IHPM, IHPMR
Chromosome Location
Chr.3: 14651683 - 14673186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016474.4 NP_057558.3 9 - 10 1044 101
NR_135523.1 8 - 9 949 101
GenBank mRNA AK091158.1 - 4 - 5 428 101
BC013999.2 - 9 - 10 1020 101
BC033897.1 - 9 - 10 1031 101

View Full Product Details