Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC28
Entrez Gene ID
123355
Gene Name
leucine rich repeat containing 28
Gene Aliases
-
Chromosome Location
Chr.15: 99251362 - 99390732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321675.1 NP_001308604.1 9 - 10 1265 70
NM_001321676.1 NP_001308605.1 10 - 11 1257 70
NM_001321677.1 NP_001308606.1 8 - 9 1062 70
NM_001321679.1 NP_001308608.1 8 - 9 1228 70
NM_001321680.1 NP_001308609.1 8 - 9 1224 70
NM_144598.4 NP_653199.2 9 - 10 1269 70
NR_135753.1 9 - 10 1174 70
NR_135754.1 9 - 10 1022 70
NR_135755.1 8 - 9 1129 70
NR_135756.1 8 - 9 1133 70
NR_135757.1 10 - 11 1243 70
NR_135758.1 9 - 10 1297 70
NR_135759.1 7 - 8 1017 70
XM_011521218.2 XP_011519520.1 9 - 10 1305 70
GenBank mRNA AK091588.1 - 8 - 9 1131 70
AK127177.1 - 4 - 5 952 70
AK292887.1 - 9 - 10 1126 70
AK295161.1 - 8 - 9 937 70
AK296352.1 - 9 - 10 1076 70
BC047306.1 - 9 - 10 1184 70
BM681658.1 - 1 - 2 70

View Full Product Details