Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD13B
Entrez Gene ID
124930
Gene Name
ankyrin repeat domain 13B
Gene Aliases
-
Chromosome Location
Chr.17: 29589769 - 29614761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152345.4 NP_689558.4 2 - 3 368 63
XM_005257920.4 XP_005257977.1 2 - 3 301 63
XM_011524319.2 XP_011522621.1 2 - 3 538 63
XM_011524320.2 XP_011522622.1 2 - 3 300 63
XM_011524321.2 XP_011522623.1 2 - 3 300 63
XM_017024174.1 XP_016879663.1 2 - 3 235 63
XM_017024175.1 XP_016879664.1 2 - 3 237 63
XM_017024176.1 XP_016879665.1 2 - 3 167 63
GenBank mRNA AK092673.1 - 2 - 3 185 63
BC032554.1 - 2 - 3 415 63
DA445860.1 - 2 - 3 235 63

View Full Product Details