The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152608.3 | NP_689821.3 | 5 - 6 | 692 | 66 |
GenBank mRNA | AK092701.1 | - | 5 - 6 | 518 | 66 |
AK127780.1 | - | 4 - 5 | 622 | 66 | |
AK291008.1 | - | 5 - 6 | 655 | 66 | |
AK308441.1 | - | 5 - 6 | 668 | 66 | |
BC071563.1 | - | 5 - 6 | 670 | 66 | |
BX648319.1 | - | 5 - 6 | 650 | 66 |