Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TOPAZ1
Entrez Gene ID
375337
Gene Name
testis and ovary specific PAZ domain containing 1
Gene Aliases
C3orf77
Chromosome Location
Chr.3: 44241886 - 44337071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145030.1 NP_001138502.1 1 - 2 510 73
XM_011533694.2 XP_011531996.1 1 - 2 510 73
XM_017006361.1 XP_016861850.1 1 - 2 510 73
XM_017006362.1 XP_016861851.1 1 - 2 510 73
GenBank mRNA AK093476.1 - 1 - 2 510 73

View Full Product Details