Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATAD2B
Entrez Gene ID
54454
Gene Name
ATPase family, AAA domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.2: 23689710 - 23927114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242338.2 NP_001229267.2 13 - 14 1916 119
NM_017552.3 NP_060022.2 13 - 14 1916 119
NR_125717.1 13 - 14 1916 119
XM_005264372.3 XP_005264429.2 14 - 15 1958 119
XM_006712030.3 XP_006712093.1 14 - 15 1958 119
XM_006712031.3 XP_006712094.1 14 - 15 1958 119
XM_011532918.2 XP_011531220.1 14 - 15 1958 119
XM_011532919.2 XP_011531221.1 14 - 15 1958 119
XM_011532920.2 XP_011531222.1 13 - 14 1916 119
XM_011532921.2 XP_011531223.1 14 - 15 1958 119
XM_011532924.2 XP_011531226.1 14 - 15 1958 119
XM_011532925.2 XP_011531227.1 14 - 15 1958 119
XM_011532928.1 XP_011531230.1 14 - 15 1958 119
XM_011532929.2 XP_011531231.1 14 - 15 1958 119
XM_011532930.2 XP_011531232.1 14 - 15 1958 119
XM_011532931.2 XP_011531233.1 14 - 15 1958 119
XM_011532932.1 XP_011531234.1 14 - 15 1958 119
XM_017004375.1 XP_016859864.1 13 - 14 1916 119
GenBank mRNA AK094821.1 - 13 - 14 1916 119
AK131301.1 - 1 - 2 56 119
BC171846.1 - 13 - 14 1863 119

View Full Product Details