The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017001115.1 | XP_016856604.1 | 14 - 15 | 10731 | 97 |
XM_017001116.1 | XP_016856605.1 | 15 - 16 | 8455 | 97 | |
XM_017001117.1 | XP_016856606.1 | 14 - 15 | 10525 | 97 | |
GenBank mRNA | AK095297.1 | - | 1 - 2 | 66 | 97 |
AL138273.1 | - | 1 - 2 | 294 | 97 |