The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011522244.1 | XP_011520546.1 | 1 - 2 | 172 | 115 |
GenBank mRNA | AK095529.1 | - | 1 - 2 | 172 | 115 |