The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_026936.1 | 1 - 2 | 483 | 129 | |
GenBank mRNA | AK096140.1 | - | 2 - 3 | 382 | 129 |
AK314737.1 | - | 1 - 2 | 483 | 129 |