The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178123.4 | NP_835224.3 | 8 - 9 | 788 | 63 |
XM_011512139.2 | XP_011510441.1 | 8 - 9 | 816 | 63 | |
XM_011512141.2 | XP_011510443.1 | 6 - 7 | 493 | 63 | |
XM_017005250.1 | XP_016860739.1 | 8 - 9 | 765 | 63 | |
XM_017005251.1 | XP_016860740.1 | 9 - 10 | 994 | 63 | |
GenBank mRNA | AK096232.1 | - | 8 - 9 | 766 | 63 |
AY209190.1 | - | 8 - 9 | 685 | 63 | |
BC047578.1 | - | 8 - 9 | 937 | 63 |