The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178171.4 | NP_835465.2 | 11 - 12 | 1211 | 92 |
XM_006721832.2 | XP_006721895.1 | 11 - 12 | 1164 | 92 | |
XM_011524651.2 | XP_011522953.1 | 9 - 10 | 927 | 92 | |
XM_017024502.1 | XP_016879991.1 | 10 - 11 | 1184 | 92 | |
GenBank mRNA | AB093591.1 | - | 10 - 11 | 1104 | 92 |
AK096439.1 | - | 12 - 13 | 1213 | 92 | |
BC109197.2 | - | 12 - 13 | 1173 | 92 |