Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD44
Entrez Gene ID
91526
Gene Name
ankyrin repeat domain 44
Gene Aliases
PP6-ARS-B
Chromosome Location
Chr.2: 196962617 - 197310797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195144.1 NP_001182073.1 3 - 4 384 70
NM_153697.2 NP_710181.2 3 - 4 384 70
XM_005246945.1 XP_005247002.1 3 - 4 374 70
XM_005246946.1 XP_005247003.1 3 - 4 374 70
XM_005246947.2 XP_005247004.1 3 - 4 374 70
XM_005246948.2 XP_005247005.1 3 - 4 374 70
XM_006712838.3 XP_006712901.1 3 - 4 374 70
XM_011512148.2 XP_011510450.1 3 - 4 374 70
XM_017005252.1 XP_016860741.1 3 - 4 374 70
GenBank mRNA AK097086.1 - 3 - 4 193 70
AK124039.1 - 3 - 4 295 70
BC016985.2 - 3 - 4 358 70
BC063622.1 - 3 - 4 325 70

View Full Product Details