The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113523.2 | NP_001106995.1 | 6 - 7 | 1065 | 107 |
NM_001308320.1 | NP_001295249.1 | 4 - 5 | 687 | 107 | |
NM_001308321.1 | NP_001295250.1 | 2 - 3 | 392 | 107 | |
NM_152615.2 | NP_689828.1 | 2 - 3 | 392 | 107 | |
XM_005247159.3 | XP_005247216.1 | 6 - 7 | 2737 | 107 | |
XM_005247160.3 | XP_005247217.1 | 5 - 6 | 2509 | 107 | |
XM_011512475.2 | XP_011510777.1 | 5 - 6 | 2621 | 107 | |
XM_011512476.2 | XP_011510778.1 | 5 - 6 | 2590 | 107 | |
XM_011512477.2 | XP_011510779.1 | 4 - 5 | 2359 | 107 | |
XM_011512478.2 | XP_011510780.1 | 6 - 7 | 848 | 107 | |
XM_011512479.2 | XP_011510781.1 | 4 - 5 | 323 | 107 | |
XM_011512480.2 | XP_011510782.1 | 6 - 7 | 2753 | 107 | |
XM_017005791.1 | XP_016861280.1 | 5 - 6 | 2509 | 107 | |
XM_017005792.1 | XP_016861281.1 | 3 - 4 | 1490 | 107 | |
GenBank mRNA | AK097515.1 | - | 8 - 9 | 1010 | 107 |
AK097916.1 | - | 2 - 3 | 329 | 107 | |
BC101701.1 | - | 2 - 3 | 392 | 107 | |
BC101703.1 | - | 2 - 3 | 392 | 107 | |
BC143573.1 | - | 2 - 3 | 392 | 107 | |
BX647656.1 | - | 4 - 5 | 663 | 107 | |
DQ063586.1 | - | 6 - 7 | 984 | 107 |