The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190274.1 | NP_001177203.1 | 22 - 23 | 2717 | 69 |
NM_025133.4 | NP_079409.3 | 22 - 23 | 2506 | 69 | |
XM_017005015.1 | XP_016860504.1 | 22 - 23 | 2664 | 69 | |
XM_017005017.1 | XP_016860506.1 | 22 - 23 | 2530 | 69 | |
GenBank mRNA | AK025477.1 | - | 7 - 8 | 709 | 69 |
AK097734.1 | - | 11 - 12 | 1254 | 69 | |
AK223592.1 | - | 22 - 23 | 2601 | 69 | |
AK292877.1 | - | 22 - 23 | 2556 | 69 | |
AK304140.1 | - | 22 - 23 | 2421 | 69 | |
AK315108.1 | - | 22 - 23 | 2506 | 69 | |
AL117620.1 | - | 5 - 6 | 488 | 69 | |
AY827075.1 | - | 22 - 23 | 2644 | 69 | |
BC012728.2 | - | 8 - 9 | 790 | 69 | |
BC043258.1 | - | 22 - 23 | 2608 | 69 | |
BC130445.1 | - | 22 - 23 | 2442 | 69 | |
BC136480.1 | - | 22 - 23 | 2442 | 69 |