The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171798.1 | NP_001165269.1 | 1 - 2 | 136 | 85 |
GenBank mRNA | AK097956.1 | - | 1 - 2 | 103 | 85 |