The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152665.2 | NP_689878.2 | 3 - 4 | 345 | 67 |
XM_011540933.2 | XP_011539235.1 | 4 - 5 | 732 | 67 | |
XM_017000566.1 | XP_016856055.1 | 3 - 4 | 641 | 67 | |
GenBank mRNA | AK098192.1 | - | 3 - 4 | 343 | 67 |
BC106885.2 | - | 3 - 4 | 310 | 67 | |
DA520308.1 | - | 3 - 4 | 345 | 67 |