The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153234.4 | NP_694966.3 | 1 - 2 | 326 | 86 |
GenBank mRNA | AK098400.1 | - | 1 - 2 | 382 | 86 |
AK291034.1 | - | 1 - 2 | 309 | 86 | |
AK307885.1 | - | 1 - 2 | 267 | 86 | |
BC036467.2 | - | 1 - 2 | 245 | 86 |