The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153234.4 | NP_694966.3 | 4 - 5 | 721 | 117 |
GenBank mRNA | AK098400.1 | - | 4 - 5 | 777 | 117 |
AK291034.1 | - | 4 - 5 | 704 | 117 | |
AK307885.1 | - | 4 - 5 | 662 | 117 | |
BC036467.2 | - | 4 - 5 | 640 | 117 |