Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP7D2
Entrez Gene ID
256714
Gene Name
MAP7 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 20006713 - 20116996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168465.1 NP_001161937.1 3 - 4 489 69
NM_001168466.1 NP_001161938.1 3 - 4 489 69
NM_001168467.1 NP_001161939.1 3 - 4 350 69
NM_152780.3 NP_689993.2 3 - 4 489 69
XM_005274478.2 XP_005274535.1 3 - 4 400 69
XM_005274479.2 XP_005274536.1 3 - 4 400 69
XM_005274480.3 XP_005274537.1 3 - 4 400 69
XM_006724481.2 XP_006724544.1 3 - 4 400 69
XM_006724482.2 XP_006724545.1 3 - 4 400 69
XM_011545483.1 XP_011543785.1 3 - 4 400 69
XM_011545484.1 XP_011543786.1 3 - 4 350 69
XM_011545485.1 XP_011543787.1 3 - 4 318 69
XM_011545486.2 XP_011543788.1 3 - 4 400 69
XM_017029393.1 XP_016884882.1 3 - 4 400 69
XM_017029394.1 XP_016884883.1 3 - 4 350 69
XM_017029395.1 XP_016884884.1 3 - 4 350 69
XM_017029396.1 XP_016884885.1 3 - 4 318 69
XM_017029397.1 XP_016884886.1 3 - 4 318 69
XM_017029398.1 XP_016884887.1 3 - 4 400 69
GenBank mRNA AK027409.1 - 3 - 4 391 69
AK098768.1 - 1 - 2 175 69
AK294788.1 - 3 - 4 344 69
AK296319.1 - 3 - 4 400 69
AK316117.1 - 3 - 4 397 69
BC037165.2 - 1 - 2 156 69
BC089400.1 - 3 - 4 410 69
BC136379.1 - 3 - 4 406 69
DA409284.1 - 3 - 4 350 69
KP849473.1 - 3 - 4 372 69
KP849474.1 - 3 - 4 372 69
KP979782.1 - 3 - 4 406 69
KP979783.1 - 3 - 4 406 69
KP979784.1 - 3 - 4 406 69
KP979785.1 - 3 - 4 406 69
KP979786.1 - 3 - 4 406 69

View Full Product Details