Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
R3HCC1
Entrez Gene ID
203069
Gene Name
R3H domain and coiled-coil containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 23288092 - 23296279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136108.2 NP_001129580.2 7 - 8 1276 70
NM_001301650.1 NP_001288579.1 8 - 9 1150 70
NR_125897.1 8 - 9 1161 70
GenBank mRNA AK293465.1 - 7 - 8 1260 70
AL050297.1 - 7 - 8 1254 70
BC032421.1 - 7 - 8 1243 70
BC050572.1 - 6 - 7 1215 70
BC143822.1 - 8 - 9 1334 70
BC143827.1 - 8 - 9 1345 70
BC143829.1 - 7 - 8 1460 70

View Full Product Details