Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARPP21
Entrez Gene ID
10777
Gene Name
cAMP regulated phosphoprotein 21
Gene Aliases
ARPP-21, R3HDM3, RCS, TARPP
Chromosome Location
Chr.3: 35638981 - 35798933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267617.1 NP_001254546.1 18 - 19 2377 92
NM_001267619.1 NP_001254548.1 18 - 19 2402 92
NM_016300.4 NP_057384.2 19 - 20 2451 92
XM_005264811.2 XP_005264868.1 20 - 21 2585 92
XM_006712943.2 XP_006713006.1 19 - 20 2483 92
XM_006712944.2 XP_006713007.1 18 - 19 2423 92
XM_011533298.2 XP_011531600.1 20 - 21 2588 92
XM_011533299.2 XP_011531601.1 20 - 21 3132 92
XM_011533300.2 XP_011531602.1 20 - 21 2751 92
XM_011533301.2 XP_011531603.1 20 - 21 2582 92
XM_011533302.2 XP_011531604.1 21 - 22 2674 92
XM_011533303.2 XP_011531605.1 21 - 22 3224 92
XM_017005574.1 XP_016861063.1 20 - 21 2678 92
XM_017005575.1 XP_016861064.1 21 - 22 14311 92
XM_017005576.1 XP_016861065.1 20 - 21 3129 92
XM_017005577.1 XP_016861066.1 20 - 21 2579 92
XM_017005578.1 XP_016861067.1 20 - 21 2555 92
XM_017005579.1 XP_016861068.1 20 - 21 3129 92
XM_017005580.1 XP_016861069.1 21 - 22 3221 92
XM_017005581.1 XP_016861070.1 21 - 22 14308 92
XM_017005582.1 XP_016861071.1 20 - 21 3126 92
XM_017005583.1 XP_016861072.1 20 - 21 2576 92
XM_017005584.1 XP_016861073.1 19 - 20 3072 92
XM_017005585.1 XP_016861074.1 19 - 20 3069 92
XM_017005586.1 XP_016861075.1 19 - 20 2525 92
XM_017005587.1 XP_016861076.1 19 - 20 2688 92
XM_017005588.1 XP_016861077.1 20 - 21 3161 92
XM_017005589.1 XP_016861078.1 20 - 21 2611 92
XM_017005590.1 XP_016861079.1 20 - 21 14248 92
XM_017005591.1 XP_016861080.1 19 - 20 3030 92
XM_017005592.1 XP_016861081.1 19 - 20 3027 92
XM_017005593.1 XP_016861082.1 19 - 20 2573 92
XM_017005594.1 XP_016861083.1 19 - 20 2477 92
XM_017005595.1 XP_016861084.1 20 - 21 3119 92
XM_017005596.1 XP_016861085.1 19 - 20 3027 92
XM_017005597.1 XP_016861086.1 19 - 20 3024 92
XM_017005598.1 XP_016861087.1 20 - 21 3116 92
XM_017005599.1 XP_016861088.1 18 - 19 2970 92
XM_017005600.1 XP_016861089.1 18 - 19 2589 92
XM_017005601.1 XP_016861090.1 18 - 19 2420 92
XM_017005602.1 XP_016861091.1 18 - 19 2967 92
XM_017005603.1 XP_016861092.1 18 - 19 2586 92
XM_017005604.1 XP_016861093.1 18 - 19 2417 92
XM_017005605.1 XP_016861094.1 19 - 20 3059 92
XM_017005606.1 XP_016861095.1 19 - 20 2509 92
XM_017005607.1 XP_016861096.1 19 - 20 2987 92
XM_017005608.1 XP_016861097.1 21 - 22 3220 92
XM_017005609.1 XP_016861098.1 21 - 22 3252 92
XM_017005610.1 XP_016861099.1 18 - 19 2918 92
XM_017005611.1 XP_016861100.1 17 - 18 2858 92
XM_017005612.1 XP_016861101.1 18 - 19 2915 92
GenBank mRNA AK294484.1 - 18 - 19 2415 92
AL133109.1 - 12 - 13 1706 92
BC031106.1 - 7 - 8 1263 92
BC036399.1 - 18 - 19 2382 92
BC051828.1 - 19 - 20 2451 92

View Full Product Details