The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005366.1 | NP_001005366.1 | 7 - 8 | 1082 | 68 |
NM_032590.4 | NP_115979.3 | 7 - 8 | 872 | 68 | |
XM_005253955.3 | XP_005254012.1 | 7 - 8 | 1082 | 68 | |
XM_005253956.3 | XP_005254013.1 | 7 - 8 | 679 | 68 | |
XM_011538867.2 | XP_011537169.1 | 7 - 8 | 2125 | 68 | |
XM_011538868.2 | XP_011537170.1 | 7 - 8 | 2121 | 68 | |
XM_011538869.2 | XP_011537171.1 | 7 - 8 | 2123 | 68 | |
GenBank mRNA | AJ459424.1 | - | 7 - 8 | 844 | 68 |
AK127328.1 | - | 7 - 8 | 1082 | 68 | |
AL133572.1 | - | 3 - 4 | 385 | 68 | |
BC115379.1 | - | 6 - 7 | 732 | 68 | |
BC115380.1 | - | 7 - 8 | 811 | 68 |