Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIK2
Entrez Gene ID
23235
Gene Name
salt inducible kinase 2
Gene Aliases
LOH11CR1I, QIK, SIK-2, SNF1LK2
Chromosome Location
Chr.11: 111602391 - 111726917 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015191.2 NP_056006.1 2 - 3 421 130
XM_017017417.1 XP_016872906.1 2 - 3 380 130
GenBank mRNA AB018324.3 - 2 - 3 322 130
AB084424.1 - 2 - 3 242 130
AB096248.1 - 2 - 3 327 130
AK000396.1 - 2 - 3 327 130
AK291233.1 - 2 - 3 356 130
AL136764.2 - 2 - 3 344 130
BC113459.1 - 2 - 3 278 130
BC117183.1 - 2 - 3 278 130
BM799630.1 - 2 - 3 375 130
DA333935.1 - 2 - 3 421 130

View Full Product Details