The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015191.2 | NP_056006.1 | 5 - 6 | 773 | 77 |
XM_017017417.1 | XP_016872906.1 | 5 - 6 | 732 | 77 | |
XM_017017418.1 | XP_016872907.1 | 3 - 4 | 302 | 77 | |
GenBank mRNA | AB018324.3 | - | 5 - 6 | 674 | 77 |
AB084424.1 | - | 5 - 6 | 594 | 77 | |
AB096248.1 | - | 5 - 6 | 679 | 77 | |
AK000396.1 | - | 5 - 6 | 679 | 77 | |
AL136764.2 | - | 5 - 6 | 696 | 77 | |
BC113459.1 | - | 5 - 6 | 630 | 77 | |
BC117183.1 | - | 5 - 6 | 630 | 77 |