The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039459.2 | NP_001034548.1 | 2 - 3 | 366 | 100 |
NM_001206480.2 | NP_001193409.1 | 17 - 18 | 2145 | 100 | |
NM_001206482.1 | NP_001193411.1 | 17 - 18 | 2315 | 100 | |
NM_014800.10 | NP_055615.8 | 17 - 18 | 1956 | 100 | |
NM_130442.3 | NP_569709.1 | 2 - 3 | 512 | 100 | |
XM_005249919.2 | XP_005249976.1 | 18 - 19 | 2509 | 100 | |
XM_006715805.1 | XP_006715868.1 | 17 - 18 | 1801 | 100 | |
XM_011515654.1 | XP_011513956.1 | 18 - 19 | 3194 | 100 | |
XM_017012838.1 | XP_016868327.1 | 18 - 19 | 3379 | 100 | |
XM_017012839.1 | XP_016868328.1 | 17 - 18 | 1743 | 100 | |
GenBank mRNA | AF398885.1 | - | 16 - 17 | 1607 | 100 |
AK126565.1 | - | 5 - 6 | 996 | 100 | |
AK291784.1 | - | 17 - 18 | 2315 | 100 | |
AK296253.1 | - | 14 - 15 | 1476 | 100 | |
AK314349.1 | - | 17 - 18 | 1745 | 100 | |
AL136787.1 | - | 17 - 18 | 1857 | 100 | |
BC003051.1 | - | 2 - 3 | 362 | 100 | |
BC064635.1 | - | 2 - 3 | 314 | 100 | |
BC077074.1 | - | 2 - 3 | 363 | 100 | |
BC114341.2 | - | 17 - 18 | 1694 | 100 | |
BI600726.1 | - | 2 - 3 | 374 | 100 | |
D87457.1 | - | 2 - 3 | 338 | 100 |