Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL23A1
Entrez Gene ID
91522
Gene Name
collagen type XXIII alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.5: 178237616 - 178590555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173465.3 NP_775736.2 28 - 29 1980 141
XM_006714933.3 XP_006714996.1 29 - 30 1788 219
XM_006714934.3 XP_006714997.1 28 - 29 1761 219
XM_006714935.3 XP_006714998.1 28 - 29 1752 219
XM_006714936.3 XP_006714999.1 28 - 29 1725 219
XM_011534688.2 XP_011532990.1 28 - 29 1761 219
XM_011534689.2 XP_011532991.1 28 - 29 1734 219
XM_011534690.2 XP_011532992.1 27 - 28 1734 219
XM_011534691.2 XP_011532993.1 25 - 26 1680 219
XM_011534692.2 XP_011532994.1 29 - 30 2813 219
GenBank mRNA AL137461.1 - 11 - 12 607 141
AY158895.1 - 28 - 29 1980 141
AY898961.1 - 28 - 29 1980 141
BC042428.1 - 29 - 30 1762 141

View Full Product Details