Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA1524
Entrez Gene ID
57650
Gene Name
KIAA1524
Gene Aliases
CIP2A, p90
Chromosome Location
Chr.3: 108545752 - 108589644 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020890.2 NP_065941.2 18 - 19 2599 106
XM_006713716.3 XP_006713779.1 18 - 19 2383 106
XM_006713717.2 XP_006713780.1 19 - 20 2495 106
XM_011513056.1 XP_011511358.1 18 - 19 2343 106
XM_011513057.2 XP_011511359.1 19 - 20 2464 106
GenBank mRNA AB040957.1 - 18 - 19 2326 106
AF334474.1 - 18 - 19 2398 106
AK022912.1 - 9 - 10 1210 106
AK056933.1 - 18 - 19 2408 106
AK308315.1 - 18 - 19 2345 106
AL713689.1 - 18 - 19 2394 106
BC017659.1 - 4 - 5 472 106
BC130564.1 - 18 - 19 2366 106
BC136371.1 - 18 - 19 2366 106

View Full Product Details