The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098475.1 | NP_001091945.1 | 8 - 9 | 1338 | 112 |
NM_182499.3 | NP_872305.3 | 8 - 9 | 1338 | 112 | |
XM_006711156.2 | XP_006711219.1 | 3 - 4 | 968 | 112 | |
XM_011509152.1 | XP_011507454.1 | 9 - 10 | 727 | 112 | |
XM_011509153.1 | XP_011507455.1 | 9 - 10 | 727 | 112 | |
XM_011509154.1 | XP_011507456.1 | 9 - 10 | 685 | 112 | |
XM_011509155.1 | XP_011507457.1 | 9 - 10 | 727 | 112 | |
XM_011509156.1 | XP_011507458.1 | 8 - 9 | 652 | 112 | |
XM_011509157.1 | XP_011507459.1 | 8 - 9 | 652 | 112 | |
XM_011509158.2 | XP_011507460.1 | 4 - 5 | 442 | 112 | |
XM_011509159.2 | XP_011507461.1 | 4 - 5 | 433 | 112 | |
XM_011509160.2 | XP_011507462.1 | 4 - 5 | 478 | 112 | |
XM_011509161.1 | XP_011507463.1 | 4 - 5 | 446 | 112 | |
XM_017000255.1 | XP_016855744.1 | 3 - 4 | 372 | 112 | |
GenBank mRNA | AK302144.1 | - | 8 - 9 | 585 | 112 |
AL713777.1 | - | 3 - 4 | 244 | 112 | |
AY358195.1 | - | 2 - 3 | 107 | 112 | |
BC044864.1 | - | 8 - 9 | 1337 | 112 | |
BC100895.1 | - | 3 - 4 | 216 | 112 | |
BC100896.1 | - | 3 - 4 | 216 | 112 | |
BC100897.2 | - | 3 - 4 | 216 | 112 | |
BC100898.2 | - | 3 - 4 | 216 | 112 |