Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMCH1
Entrez Gene ID
22998
Gene Name
LIM and calponin homology domains 1
Gene Aliases
LIMCH1A, LMO7B
Chromosome Location
Chr.4: 41359607 - 41700044 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112717.2 NP_001106188.1 6 - 7 539 86
NM_001112718.2 NP_001106189.1 6 - 7 539 86
NM_001289122.1 NP_001276051.1 6 - 7 539 86
NM_001289124.1 NP_001276053.1 6 - 7 584 86
NM_014988.3 NP_055803.2 6 - 7 539 86
XM_005248057.1 XP_005248114.1 6 - 7 539 86
XM_005248058.1 XP_005248115.1 6 - 7 539 86
XM_005248060.1 XP_005248117.1 6 - 7 539 86
XM_005248061.1 XP_005248118.1 6 - 7 539 86
XM_005248067.2 XP_005248124.1 4 - 5 1413 86
XM_005248072.1 XP_005248129.1 6 - 7 539 86
XM_005248074.1 XP_005248131.1 6 - 7 539 86
XM_005248075.1 XP_005248132.1 6 - 7 539 86
XM_006713996.1 XP_006714059.1 6 - 7 539 86
XM_011513642.1 XP_011511944.1 6 - 7 539 86
XM_011513643.1 XP_011511945.1 6 - 7 539 86
XM_011513644.1 XP_011511946.1 6 - 7 539 86
XM_011513645.1 XP_011511947.1 6 - 7 539 86
XM_011513646.1 XP_011511948.1 6 - 7 539 86
XM_011513648.1 XP_011511950.1 6 - 7 539 86
XM_011513651.2 XP_011511953.1 4 - 5 1418 86
XM_011513653.1 XP_011511955.1 6 - 7 539 86
XM_011513654.1 XP_011511956.1 6 - 7 539 86
XM_011513658.2 XP_011511960.1 6 - 7 800 86
XM_017007894.1 XP_016863383.1 6 - 7 539 86
XM_017007895.1 XP_016863384.1 6 - 7 539 86
XM_017007896.1 XP_016863385.1 6 - 7 539 86
XM_017007897.1 XP_016863386.1 6 - 7 539 86
XM_017007898.1 XP_016863387.1 6 - 7 539 86
XM_017007899.1 XP_016863388.1 6 - 7 539 86
XM_017007900.1 XP_016863389.1 6 - 7 539 86
XM_017007901.1 XP_016863390.1 6 - 7 539 86
XM_017007902.1 XP_016863391.1 6 - 7 539 86
GenBank mRNA AB029025.2 - 6 - 7 537 86
AL831962.2 - 4 - 5 294 86
BC023546.2 - 3 - 4 392 86
BC053639.1 - 5 - 6 622 86
BC095394.1 - 6 - 7 415 86
CR933645.1 - 6 - 7 540 86
CR936601.1 - 6 - 7 510 86
CR936610.1 - 6 - 7 585 86
CR936658.1 - 6 - 7 518 86
CR936661.1 - 6 - 7 479 86
CR936664.1 - 6 - 7 540 86

View Full Product Details