Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMCH1
Entrez Gene ID
22998
Gene Name
LIM and calponin homology domains 1
Gene Aliases
LIMCH1A, LMO7B
Chromosome Location
Chr.4: 41359607 - 41700044 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112717.2 NP_001106188.1 7 - 8 734 74
NM_001112718.2 NP_001106189.1 7 - 8 734 74
NM_001112719.2 NP_001106190.1 2 - 3 519 74
NM_001112720.2 NP_001106191.1 2 - 3 519 74
NM_001289122.1 NP_001276051.1 7 - 8 734 74
NM_001289124.1 NP_001276053.1 7 - 8 779 74
NM_014988.3 NP_055803.2 7 - 8 734 74
XM_005248057.1 XP_005248114.1 7 - 8 734 74
XM_005248058.1 XP_005248115.1 7 - 8 734 74
XM_005248060.1 XP_005248117.1 7 - 8 734 74
XM_005248061.1 XP_005248118.1 7 - 8 734 74
XM_005248067.2 XP_005248124.1 5 - 6 1608 74
XM_005248072.1 XP_005248129.1 7 - 8 734 74
XM_005248074.1 XP_005248131.1 7 - 8 734 74
XM_005248075.1 XP_005248132.1 7 - 8 734 74
XM_006713996.1 XP_006714059.1 7 - 8 734 74
XM_011513642.1 XP_011511944.1 7 - 8 734 74
XM_011513643.1 XP_011511945.1 7 - 8 734 74
XM_011513644.1 XP_011511946.1 7 - 8 734 74
XM_011513645.1 XP_011511947.1 7 - 8 734 74
XM_011513646.1 XP_011511948.1 7 - 8 734 74
XM_011513648.1 XP_011511950.1 7 - 8 734 74
XM_011513649.1 XP_011511951.1 2 - 3 393 74
XM_011513651.2 XP_011511953.1 5 - 6 1613 74
XM_011513653.1 XP_011511955.1 7 - 8 734 74
XM_011513654.1 XP_011511956.1 7 - 8 734 74
XM_011513655.1 XP_011511957.1 2 - 3 393 74
XM_011513656.1 XP_011511958.1 2 - 3 393 74
XM_011513657.1 XP_011511959.1 2 - 3 393 74
XM_011513658.2 XP_011511960.1 7 - 8 995 74
XM_017007894.1 XP_016863383.1 7 - 8 734 74
XM_017007895.1 XP_016863384.1 7 - 8 734 74
XM_017007896.1 XP_016863385.1 7 - 8 734 74
XM_017007897.1 XP_016863386.1 7 - 8 734 74
XM_017007898.1 XP_016863387.1 7 - 8 734 74
XM_017007899.1 XP_016863388.1 7 - 8 734 74
XM_017007900.1 XP_016863389.1 7 - 8 734 74
XM_017007901.1 XP_016863390.1 7 - 8 734 74
XM_017007902.1 XP_016863391.1 7 - 8 734 74
XM_017007903.1 XP_016863392.1 2 - 3 393 74
XM_017007904.1 XP_016863393.1 2 - 3 393 74
XM_017007905.1 XP_016863394.1 2 - 3 393 74
GenBank mRNA AB029025.2 - 7 - 8 732 74
AK294774.1 - 2 - 3 582 74
AK295784.1 - 1 - 2 108 74
AK295836.1 - 1 - 2 182 74
AK299297.1 - 2 - 3 389 74
AK299835.1 - 2 - 3 254 74
AK309846.1 - 2 - 3 461 74
AL831962.2 - 5 - 6 489 74
BC023546.2 - 4 - 5 587 74
BC053639.1 - 6 - 7 817 74
BC068200.1 - 2 - 3 400 74
BC095394.1 - 7 - 8 610 74
BX537916.1 - 2 - 3 389 74
BX640692.1 - 1 - 2 149 74
CR749205.1 - 2 - 3 389 74
CR933645.1 - 7 - 8 735 74
CR936601.1 - 7 - 8 705 74
CR936610.1 - 7 - 8 780 74
CR936658.1 - 7 - 8 713 74
CR936661.1 - 7 - 8 674 74
CR936664.1 - 7 - 8 735 74
DA786971.1 - 2 - 3 395 74

View Full Product Details