The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152716.2 | NP_689929.2 | 18 - 19 | 2430 | 82 |
XM_005273833.4 | XP_005273890.1 | 18 - 19 | 3552 | 82 | |
XM_005273834.1 | XP_005273891.1 | 17 - 18 | 2342 | 82 | |
XM_011544811.2 | XP_011543113.1 | 14 - 15 | 1765 | 82 | |
XM_017017339.1 | XP_016872828.1 | 19 - 20 | 2506 | 82 | |
GenBank mRNA | AB065087.1 | - | 3 - 4 | 329 | 82 |
AK094193.1 | - | 15 - 16 | 1941 | 82 | |
AK127649.1 | - | 7 - 8 | 872 | 82 | |
AK127943.1 | - | 17 - 18 | 2189 | 82 | |
AL831992.2 | - | 18 - 19 | 2371 | 82 | |
BC048309.1 | - | 4 - 5 | 413 | 82 | |
BC065264.1 | - | 15 - 16 | 1924 | 82 | |
BC109038.1 | - | 15 - 16 | 1885 | 82 | |
BC109039.1 | - | 15 - 16 | 1885 | 82 | |
BC111047.1 | - | 16 - 17 | 2092 | 82 |