The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277813.1 | NP_001264742.1 | 5 - 6 | 2199 | 118 |
NM_001277814.1 | NP_001264743.1 | 5 - 6 | 1953 | 118 | |
NM_001277815.1 | NP_001264744.1 | 5 - 6 | 2084 | 118 | |
NM_001277816.1 | NP_001264745.1 | 3 - 4 | 1967 | 118 | |
NM_001277817.1 | NP_001264746.1 | 4 - 5 | 1892 | 118 | |
NM_001277818.1 | NP_001264747.1 | 4 - 5 | 1813 | 118 | |
NM_152758.5 | NP_689971.4 | 4 - 5 | 2053 | 118 | |
XM_011517509.2 | XP_011515811.1 | 5 - 6 | 2046 | 118 | |
XM_011517510.2 | XP_011515812.1 | 3 - 4 | 1809 | 118 | |
GenBank mRNA | AK093081.1 | - | 4 - 5 | 1813 | 118 |
AK127574.1 | - | 4 - 5 | 1985 | 118 | |
AL831816.1 | - | 4 - 5 | 1888 | 118 | |
AL832005.1 | - | 4 - 5 | 1986 | 118 | |
AL832335.1 | - | 5 - 6 | 1981 | 118 | |
AL832369.1 | - | 2 - 3 | 1687 | 118 | |
BC052970.1 | - | 4 - 5 | 2002 | 118 | |
BX537996.2 | - | 4 - 5 | 1990 | 118 |