Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC30
Entrez Gene ID
728621
Gene Name
coiled-coil domain containing 30
Gene Aliases
PFD6L, PFDN6L
Chromosome Location
Chr.1: 42521240 - 42654664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080850.2 NP_001074319.1 4 - 5 535 73
XM_011542067.2 XP_011540369.1 5 - 6 1467 73
XM_011542068.2 XP_011540370.1 5 - 6 1472 73
XM_011542069.2 XP_011540371.1 4 - 5 1326 73
XM_011542070.2 XP_011540372.1 3 - 4 493 73
XM_017002225.1 XP_016857714.1 4 - 5 552 73
XM_017002226.1 XP_016857715.1 5 - 6 863 73
GenBank mRNA AK302075.1 - 6 - 7 719 73
AL832217.1 - 10 - 11 1430 73

View Full Product Details