The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256054.2 | NP_001242983.1 | 4 - 5 | 799 | 147 |
NM_018325.4 | NP_060795.1 | 4 - 5 | 704 | 147 | |
GenBank mRNA | AK291425.1 | - | 4 - 5 | 694 | 147 |
AL832229.1 | - | 3 - 4 | 661 | 147 | |
AL832467.1 | - | 4 - 5 | 660 | 147 | |
BC068445.1 | - | 3 - 4 | 653 | 147 | |
JN681271.1 | - | 4 - 5 | 777 | 147 |