Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK8
Entrez Gene ID
81704
Gene Name
dedicator of cytokinesis 8
Gene Aliases
HEL-205, MRD2, ZIR8
Chromosome Location
Chr.9: 211297 - 465260 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190458.1 NP_001177387.1 19 - 20 2338 63
NM_001193536.1 NP_001180465.1 19 - 20 2338 63
NM_203447.3 NP_982272.2 20 - 21 2553 63
XM_011518045.2 XP_011516347.1 20 - 21 2408 63
XM_011518046.2 XP_011516348.1 20 - 21 2451 63
XM_011518047.2 XP_011516349.1 20 - 21 2634 63
XM_011518048.2 XP_011516350.1 19 - 20 2373 63
XM_011518049.2 XP_011516351.1 7 - 8 1113 63
XM_017015173.1 XP_016870662.1 20 - 21 2408 63
XM_017015174.1 XP_016870663.1 20 - 21 2488 63
GenBank mRNA AB191037.1 - 20 - 21 2553 63
AK074081.1 - 6 - 7 671 63
AK090429.1 - 12 - 13 1542 63
AL832270.1 - 18 - 19 4394 63
AY117695.1 - 7 - 8 1229 63
BC045629.1 - 21 - 22 3515 63
BC130518.1 - 20 - 21 2316 63
BC143929.1 - 20 - 21 2316 63
HM991480.1 - 19 - 20 2334 63

View Full Product Details