Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- OBSCN
- Entrez Gene ID
-
84033
- Gene Name
- obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
- Gene Aliases
-
ARHGEF30, UNC89
- Chromosome Location
-
Chr.1: 228208013 - 228381431 on Build GRCh38
View Full Product Details